Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257477.2 | XP_006257539.1 | 5 | 584 | ||
XM_006257478.2 | XP_006257540.1 | 4 | 523 | |||
XM_008773510.1 | XP_008771732.1 | 5 | 638 |
Gene Symbol: Rhox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135607.1 | NP_001129079.1 | 837 | |||
XM_008773382.1 | XP_008771604.1 | 638 | ||||
XM_008773383.1 | XP_008771605.1 | 5 | 584 | |||
XM_008773384.1 | XP_008771606.1 | 523 | ||||
GenBank mRNA | DQ058651.1 | - | 837 |