Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135878.1 | NP_001129350.1 | 32 | 5487 | ||
XM_006229587.2 | XP_006229649.1 | 33 | 5645 | |||
XM_008773630.1 | XP_008771852.1 | 33 | 5699 | |||
XM_008773631.1 | XP_008771853.1 | 34 | 5906 | |||
XM_008773632.1 | XP_008771854.1 | 32 | 5537 |