Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139507.1 | NP_001132979.1 | 3 | 1481 | ||
XM_006229593.2 | XP_006229655.1 | 3 | 1460 | |||
XM_006229594.2 | XP_006229656.1 | 3 | 1459 | |||
XM_006229596.2 | XP_006229658.1 | 3 | 1457 | |||
XM_006229597.1 | XP_006229659.1 | 3 | 1476 |