Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248404.2 | XP_006248466.1 | 4 | 1596 | ||
XM_344023.7 | XP_344024.5 | 3 | 1481 | |||
GenBank mRNA | FQ217730.1 | - | 1621 | |||
FQ223618.1 | - | 1621 |