Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025751.1 | NP_001020922.1 | 7 | 856 | ||
XM_006252188.2 | XP_006252250.1 | 8 | 1250 | |||
XM_008770769.1 | XP_008768991.1 | 7 | 1277 | |||
XM_008770770.1 | XP_008768992.1 | 6 | 516 | |||
GenBank mRNA | BC098683.1 | - | 7 | 856 | 7385175 |