Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228711.2 | XP_006228773.1 | 4 | 1483 | ||
XM_006228712.2 | XP_006228774.1 | 4 | 1478 |
Gene Symbol: Zfp74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271339.1 | NP_001258268.1 | 3 | 334 | ||
XM_006228731.2 | XP_006228793.1 | 4 | 1483 | |||
XM_006228732.2 | XP_006228794.1 | 4 | 1478 |