Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108915.1 | NP_001102385.1 | 6 | 1633 | ||
XM_006229349.2 | XP_006229411.1 | 7 | 1835 | |||
XM_006229350.2 | XP_006229412.1 | 7 | 1801 | |||
XM_008759545.1 | XP_008757767.1 | 6 | 2779 |