Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191896.1 | NP_001178825.1 | 15 | 2297 | ||
XM_008760160.1 | XP_008758382.1 | 17 | 4492 |