Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108937.1 | NP_001102407.1 | 2 | 199 | ||
XM_008760778.1 | XP_008759000.1 | 2 | 497 | |||
XM_008760779.1 | XP_008759001.1 | 2 | 256 | |||
XM_008760780.1 | XP_008759002.1 | 2 | 478 | |||
GenBank mRNA | BC166996.1 | - | 2 | 70 |