Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108941.1 | NP_001102411.1 | 13, 14 | 1472 | ||
XM_006232190.2 | XP_006232252.1 | 12, 13 | 1365 | |||
XM_008760889.1 | XP_008759111.1 | 13, 14 | 2228 |