Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014253.2 | NP_001014275.2 | 3 | 316 | ||
GenBank mRNA | AY995234.1 | - | 3 | 316 | ||
BC086953.1 | - | 3 | 210 | 7321890 |