Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233100.2 | XP_006233162.1 | 12 | 2169 | ||
XM_345266.7 | XP_345267.3 | 12 | 2169 |