Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108958.1 | NP_001102428.1 | 10 | 1264 | ||
XM_006234783.1 | XP_006234845.1 | 11 | 1411 |