Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001067558.5 | XP_001067558.2 | 1 | 1191 | ||
XM_008764541.1 | XP_008762763.1 | 1 | 1198 |