Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239960.2 | XP_006240022.1 | 1 | 213 | ||
XM_006239962.2 | XP_006240024.1 | 1 | 212 |