Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEWGENE_1310847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764977.1 | XP_008763199.1 | 1 | 882 | ||
XM_008764978.1 | XP_008763200.1 | 2 | 1051 | |||
XM_008764979.1 | XP_008763201.1 | 2 | 1067 | |||
XM_008764980.1 | XP_008763202.1 | 2 | 1069 |
Gene Symbol: Bag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008526.2 | NP_001008526.1 | 993 | |||
XM_006240601.2 | XP_006240663.1 | 1153 | ||||
XM_006240602.2 | XP_006240664.1 | 1093 | ||||
XM_006240603.2 | XP_006240665.1 | 1069 | ||||
GenBank mRNA | AY366364.1 | - | 1038 | |||
BC088418.1 | - | 993 | 7123242 |