Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079943.1 | NP_001073412.1 | 8 | 824 | ||
XM_006242666.1 | XP_006242728.1 | 7 | 831 | |||
XM_006242667.2 | XP_006242729.1 | 7 | 2919 | |||
XM_006242668.2 | XP_006242730.1 | 8 | 2225 | |||
XM_006242670.1 | XP_006242732.1 | 8 | 914 | |||
XM_006242671.2 | XP_006242733.1 | 6 | 700 | |||
XM_006242672.2 | XP_006242734.1 | 7 | 2427 | |||
XM_006242673.2 | XP_006242735.1 | 6 | 2280 | |||
XM_008765981.1 | XP_008764203.1 | 7 | 2430 | |||
XM_008765982.1 | XP_008764204.1 | 7 | 767 | |||
XM_008765983.1 | XP_008764205.1 | 7 | 2075 | |||
XM_008765984.1 | XP_008764206.1 | 7 | 2031 | |||
GenBank mRNA | BC129125.1 | - | 8 | 824 | 7133611 |