Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001934.1 | NP_001001934.1 | 3 | 294 | ||
XM_008772747.1 | XP_008770969.1 | 4 | 981 | |||
GenBank mRNA | HE864428.1 | - | 2 | 205 | ||
HE864429.1 | - | 3 | 334 |
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HE864431.1 | - | 577 | |||
HE864432.1 | - | 1726 | ||||
HE864433.1 | - | 1951 | ||||
HE864434.1 | - | 2207 |