Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031608.1 | NP_113796.1 | 14 | 2677 | ||
GenBank mRNA | M36418.2 | - | 14 | 2480 | ||
M38060.1 | - | 14 | 2480 | |||
X17184.1 | - | 14 | 2677 |