Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019305.2 | NP_062178.1 | 3 | 914 | ||
GenBank mRNA | M22427.1 | - | 914 | |||
X07285.1 | - | 3 | 635 |