Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031632.1 | NP_113820.1 | 9 | 1358 | ||
GenBank mRNA | AF262319.1 | - | 9 | 1358 | ||
AF271996.1 | - | 9 | 1325 | |||
AF286006.1 | - | 9 | 1358 | |||
AF308469.1 | - | 9 | 1325 | |||
AY027667.1 | - | 9 | 1334 |