Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021754.1 | NP_068522.1 | 9 | 1010 | ||
XM_006245013.1 | XP_006245075.1 | 8 | 835 | |||
GenBank mRNA | AF194371.1 | - | 9 | 1010 | ||
BC087637.1 | - | 9 | 1043 | 7309062 | ||
FQ225580.1 | - | 2 | 206 | |||
FQ230388.1 | - | 2 | 206 |