Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021767.2 | NP_068535.2 | 1880 | |||
GenBank mRNA | M96374.1 | - | 1880 |