Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021771.2 | NP_068539.2 | 5 | 1400 | ||
XM_006232282.2 | XP_006232344.2 | 5 | 1565 | |||
XM_008760941.1 | XP_008759163.1 | 5 | 1565 | |||
GenBank mRNA | JN160741.1 | - | 5 | 1161 |