Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021848.1 | NP_068620.1 | 5 | 585 | ||
GenBank mRNA | AF105368.1 | - | 5 | 585 |
Set Membership: |
G-protein coupled receptor |