Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021853.1 | NP_068625.1 | 27 | 2989 | ||
XM_006233697.2 | XP_006233759.1 | 27 | 3303 | |||
XM_006233698.2 | XP_006233760.1 | 27 | 3297 | |||
XM_006233699.2 | XP_006233761.1 | 27 | 3303 | |||
XM_006233700.2 | XP_006233762.1 | 26 | 3261 | |||
XM_006233701.2 | XP_006233763.1 | 27 | 3303 | |||
XM_006233702.2 | XP_006233764.1 | 26 | 3261 | |||
XM_006233703.2 | XP_006233765.1 | 27 | 3303 | |||
XM_006233704.2 | XP_006233766.1 | 26 | 3183 | |||
XM_006233705.2 | XP_006233767.1 | 26 | 3174 | |||
XM_006233706.2 | XP_006233768.1 | 27 | 3303 | |||
XM_006233707.2 | XP_006233769.1 | 26 | 3178 | |||
XM_008761624.1 | XP_008759846.1 | 27 | 3293 | |||
GenBank mRNA | AF089730.1 | - | 27 | 2989 | ||
AY884213.1 | - | 26 | 2887 |