Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764322.1 | XP_008762544.1 | 13 | 2175 | ||
GenBank mRNA | X59290.1 | - | 3 | 231 |