Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031682.1 | NP_113870.1 | 5 | 564 | ||
GenBank mRNA | AF049878.1 | - | 5 | 552 | ||
AF069770.1 | - | 5 | 564 | |||
BC158587.1 | - | 5 | 558 | 9089087 | ||
FQ210583.1 | - | 3 | 305 | |||
FQ220319.1 | - | 3 | 305 | |||
FQ225948.1 | - | 3 | 305 | |||
FQ228203.1 | - | 3 | 305 | |||
FQ230672.1 | - | 3 | 306 | |||
FQ231826.1 | - | 3 | 305 | |||
FQ232016.1 | - | 3 | 305 |