Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270692.1 | NP_001257621.1 | 6 | 503 | ||
NM_001270693.1 | NP_001257622.1 | 6 | 503 | |||
NM_001270694.1 | NP_001257623.1 | 6 | 503 | |||
NM_022244.2 | NP_071580.1 | 6 | 503 | |||
XM_006230194.2 | XP_006230256.1 | 6 | 555 | |||
XM_008759791.1 | XP_008758013.1 | 6 | 555 | |||
XM_008759792.1 | XP_008758014.1 | 6 | 556 | |||
XM_008759793.1 | XP_008758015.1 | 6 | 555 | |||
XM_008759794.1 | XP_008758016.1 | 6 | 554 | |||
GenBank mRNA | AB042827.1 | - | 6 | 502 | ||
AB060556.1 | - | 6 | 502 | |||
AB060557.1 | - | 6 | 502 | |||
AB080637.1 | - | 6 | 502 | |||
BC085736.1 | - | 6 | 504 | 7130218 |