Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022254.3 | NP_071590.1 | 2 | 773 | ||
XM_006236124.2 | XP_006236186.1 | 4 | 1712 | |||
XM_006236126.2 | XP_006236188.1 | 3 | 1588 | |||
XM_008762724.1 | XP_008760946.1 | 3 | 1503 | |||
GenBank mRNA | AB040803.1 | - | 1 | 511 | ||
AF203907.1 | - | 1 | 535 | |||
BC087727.1 | - | 2 | 773 | 7320496 |
Set Membership: |
G-protein coupled receptor |