Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mre11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022279.1 | NP_071615.1 | 14 | 1637 | ||
XM_006242532.2 | XP_006242594.1 | 14 | 1732 | |||
XM_006242533.1 | XP_006242595.1 | 14 | 1726 | |||
XM_006242534.2 | XP_006242596.1 | 15 | 1808 | |||
GenBank mRNA | AF218574.1 | - | 14 | 1637 |