Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130894.4 | NP_570964.4 | 17, 18 | 2356 | ||
XM_006239402.2 | XP_006239464.1 | 18, 19 | 2509 | |||
XM_006239403.1 | XP_006239465.1 | 18, 19 | 2538 | |||
XM_008764288.1 | XP_008762510.1 | 19, 20 | 2526 | |||
GenBank mRNA | AB084165.1 | - | 16, 17 | 2209 | ||
BC070880.1 | - | 17, 18 | 2360 | 7096620 | ||
U41803.2 | - | 18, 19 | 2374 |
Gene Symbol: LOC100911485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750086.3 | XP_003750134.1 | 2829 |