Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270838.1 | NP_001257767.1 | 4 | 480 | ||
NM_001270839.1 | NP_001257768.1 | 4 | 480 | |||
NM_001270840.1 | NP_001257769.1 | 4 | 480 | |||
NM_022941.4 | NP_075230.1 | 4 | 480 | |||
XM_006250261.2 | XP_006250323.1 | 4 | 589 | |||
XM_008769758.1 | XP_008767980.1 | 4 | 592 | |||
XM_008769759.1 | XP_008767981.1 | 4 | 592 | |||
GenBank mRNA | AB104736.1 | - | 4 | 533 | ||
AB105072.1 | - | 4 | 533 | |||
AB204897.1 | - | 3 | 331 | |||
AB204898.1 | - | 3 | 331 | |||
AB204899.1 | - | 3 | 331 | |||
AB204900.1 | - | 3 | 331 | |||
AF133094.1 | - | 3 | 331 | |||
AF133095.1 | - | 3 | 331 | |||
BC089763.1 | - | 4 | 480 | 7324318 |