Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022950.1 | NP_075239.1 | 4 | 1126 | ||
XM_006236091.2 | XP_006236153.1 | 5 | 1147 | |||
XM_008762725.1 | XP_008760947.1 | 5 | 1096 | |||
GenBank mRNA | AF157963.1 | - | 4 | 1126 |