Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023022.2 | NP_075411.2 | 5 | 766 | ||
GenBank mRNA | AB015194.1 | - | 5 | 739 | ||
AF531097.1 | - | 5 | 766 |