Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023959.1 | NP_076449.1 | 9 | 2261 | ||
GenBank mRNA | AB042272.1 | - | 6 | 1118 |