Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270855.1 | NP_001257784.1 | 6 | 708 | ||
NM_001270856.1 | NP_001257785.1 | 5 | 627 | |||
NM_001270857.1 | NP_001257786.1 | 5 | 624 | |||
NM_001270858.1 | NP_001257787.1 | 5 | 627 | |||
NM_001270859.1 | NP_001257788.1 | 6 | 639 | |||
NM_019176.2 | NP_062049.1 | 6 | 708 | |||
XM_006252149.2 | XP_006252211.1 | 7 | 862 | |||
XM_006252151.2 | XP_006252213.1 | 6 | 1771 | |||
XM_006252156.2 | XP_006252218.1 | 5 | 1683 | |||
XM_008770778.1 | XP_008769000.1 | 6 | 712 | |||
GenBank mRNA | AF026528.1 | - | 5 | 568 | ||
AF026529.1 | - | 5 | 568 | |||
AF026530.1 | - | 6 | 649 | |||
BC092646.1 | - | 6 | 645 | 7319130 | ||
CB580507.1 | - | 5 | 558 | |||
FQ212720.1 | - | 4 | 438 | |||
FQ212976.1 | - | 5 | 578 | |||
FQ213134.1 | - | 5 | 628 |