Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173103.1 | NP_775126.1 | 14 | 1562 | ||
XM_006239320.2 | XP_006239382.1 | 17 | 4434 | |||
XM_008764293.1 | XP_008762515.1 | 16 | 4102 | |||
XM_008764294.1 | XP_008762516.1 | 16 | 2368 | |||
XM_008764295.1 | XP_008762517.1 | 15 | 2513 | |||
XM_008764296.1 | XP_008762518.1 | 14 | 2391 | |||
XM_008764297.1 | XP_008762519.1 | 14 | 2373 | |||
GenBank mRNA | BC081698.1 | - | 14 | 2057 | ||
D26111.1 | - | 15 | 1616 | |||
DQ437671.1 | - | 15 | 1666 | |||
DQ437672.1 | - | 15 | 1559 | |||
DQ437673.1 | - | 14 | 1537 | |||
DQ437674.1 | - | 14 | 1501 | |||
DQ437675.1 | - | 15 | 1509 | |||
DQ437676.1 | - | 14 | 1487 | |||
Z30663.1 | - | 14 | 1562 |