Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031753.1 | NP_113941.1 | 3, 4 | 581 | ||
GenBank mRNA | AB008538.1 | - | 3, 4 | 581 | ||
BC061970.1 | - | 3, 4 | 590 | 5621856 | ||
Y13240.1 | - | 1, 2 | 114 |