Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc21a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030837.1 | NP_110464.1 | 10 | 1166 | ||
GenBank mRNA | AB012662.1 | - | 9 | 1055 | ||
AF445993.1 | - | 8 | 1008 | |||
AF445994.1 | - | 8 | 961 | |||
AF445996.1 | - | 8 | 833 | |||
AF445997.1 | - | 9 | 1094 | |||
AF446000.1 | - | 6 | 733 | |||
AF446001.1 | - | 6 | 759 | |||
AF446005.1 | - | 6 | 605 | |||
AF446006.1 | - | 7 | 866 | |||
D79981.1 | - | 10 | 1166 |