Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc21a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030837.1 | NP_110464.1 | 11 | 1346 | ||
GenBank mRNA | AB012662.1 | - | 10 | 1235 | ||
AF445993.1 | - | 9 | 1188 | |||
AF445994.1 | - | 9 | 1141 | |||
AF445995.1 | - | 9 | 1109 | |||
AF445996.1 | - | 9 | 1013 | |||
AF445997.1 | - | 10 | 1274 | |||
AF445998.1 | - | 8 | 1023 | |||
AF445999.1 | - | 7 | 890 | |||
AF446000.1 | - | 7 | 913 | |||
AF446001.1 | - | 7 | 939 | |||
AF446002.1 | - | 7 | 881 | |||
AF446003.1 | - | 6 | 748 | |||
AF446004.1 | - | 6 | 732 | |||
AF446005.1 | - | 7 | 785 | |||
AF446006.1 | - | 8 | 1046 | |||
BC085730.1 | - | 4 | 428 | 7110926 | ||
D79981.1 | - | 11 | 1346 |