Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031010.2 | NP_112272.2 | 8 | 1108 | ||
XM_006246823.2 | XP_006246885.1 | 10 | 1389 | |||
XM_006246824.2 | XP_006246886.1 | 9 | 1284 | |||
GenBank mRNA | FQ233819.1 | - | 2 | 236 | ||
S69383.1 | - | 8 | 1108 |