Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035238.2 | NP_001030315.1 | 7 | 700 | ||
NM_031021.3 | NP_112283.1 | 7 | 751 | |||
XM_006256081.2 | XP_006256143.1 | 7 | 787 | |||
GenBank mRNA | BC078807.1 | - | 7 | 700 | 7133064 | |
EV778558.1 | - | 7 | 754 | |||
FQ214437.1 | - | 7 | 767 | |||
FQ230013.1 | - | 7 | 739 | |||
HE864425.1 | - | 3 | 253 | |||
HE864426.1 | - | 6 | 635 | |||
HE864432.1 | - | 635 | ||||
HE864433.1 | - | 635 | ||||
HE864434.1 | - | 635 |