Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031043.2 | NP_112305.1 | 3 | 418 | ||
GenBank mRNA | AF021343.1 | - | 3 | 421 | ||
BC070944.1 | - | 4 | 459 | 7103004 | ||
FQ215156.1 | - | 1 | 37 | |||
U96130.1 | - | 2 | 155 |