Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053334.1 | NP_445786.1 | 4 | 915 | ||
XM_006253658.2 | XP_006253720.1 | 4 | 957 | |||
XM_006253659.2 | XP_006253721.1 | 3 | 892 | |||
GenBank mRNA | AF302085.1 | - | 4 | 915 | ||
BC076377.1 | - | 4 | 917 | 7110159 |