Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052809.1 | NP_434696.1 | 2, 3 | 438 | ||
XM_006254698.2 | XP_006254760.1 | 2, 3 | 663 | |||
GenBank mRNA | BC070509.1 | - | 2, 3 | 429 | 7100321 | |
FQ218376.1 | - | 1, 2 | 75 | |||
FQ219282.1 | - | 1, 2 | 75 | |||
M35266.1 | - | 2, 3 | 438 |