Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270621.1 | NP_001257550.1 | 2 | 355 | ||
NM_031075.2 | NP_112337.2 | 2 | 355 | |||
GenBank mRNA | AF084975.1 | - | 2 | 200 | ||
BC081783.1 | - | 3 | 383 | 7134300 | ||
X90651.1 | - | 2 | 355 | |||
X91167.1 | - | 2 | 345 |