Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031086.2 | NP_112348.2 | 13 | 1650 | ||
XM_006240719.2 | XP_006240781.1 | 11 | 1936 | |||
XM_008765045.1 | XP_008763267.1 | 11 | 1382 | |||
GenBank mRNA | CK366505.1 | - | 1 | 75 | ||
CV105055.1 | - | 4 | 605 |