Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031119.2 | NP_112381.2 | 3 | 189 | ||
XM_006234321.2 | XP_006234383.1 | 3 | 265 | |||
XM_006234322.2 | XP_006234384.1 | 3 | 265 | |||
XM_006234323.2 | XP_006234385.1 | 3 | 265 | |||
GenBank mRNA | BC081780.1 | - | 3 | 189 | 7192454 | |
BC097252.1 | - | 3 | 195 | |||
X67859.1 | - | 2 | 124 |