Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031149.1 | NP_112411.1 | 8 | 711 | ||
GenBank mRNA | AB000491.1 | - | 8 | 711 | ||
BC058462.1 | - | 8 | 707 | 6891005 | ||
D83521.1 | - | 8 | 729 | |||
FQ213263.1 | - | 8 | 728 | |||
FQ213735.1 | - | 8 | 703 | |||
FQ233831.1 | - | 8 | 714 |